Learn More
CD30 Mouse anti-Human, Brilliant Violet 421, Clone: BerH8, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 562876
Description
The BerH8 monoclonal antibody specifically binds to CD30. CD30 is a 120 kDa type I transmembrane glycoprotein that is expressed on stimulated T and B cells. CD30 is an activation marker which that initially identified to be expressed on Reed-Sternberg cells. It is also expressed on a few extrafollicular T and B cells located at the rim of germinal centers. CD30 serves as a cytokine receptor. It belongs to the nerve growth factor receptor/tumor necrosis factor receptor (NGFR/TNFR) superfamily and is also known as TNFRSF8. CD30 interaction with CD30 ligand (CD30L/CD153/TNFSF8) can mediate signals for proliferation, apoptosis and cytotoxicity of lymphoid cells.
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon™ BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon™ BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue™ conjugates.
Specifications
| CD30 | |
| Monoclonal | |
| Brilliant Violet 421 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| BerH8 | |
| TNFRSF8; TNR8; CD30L/CD153 receptor; Ki-1; D1S166E | |
| Mouse | |
| 100 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.