Learn More
CD3 Mouse anti-Rat, PE, Clone: 1F4, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 550353
Description
The 1F4 antibody reacts with the T-cell receptor-associated CD3 cell-surface antigen found on thymocytes, peripheral T lymphocytes, and dendritic epidermal T cells. In vitro, immobilized 1F4 mAb induces T-cell proliferation, and soluble mAb synergizes with PMA for T-cell activation. Pre-incubation of splenocytes with mAb 1F4 blocks staining with mAb G4.18 (Cat. No. 559974/554830).
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
| CD3 | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| F344 rat splenic T cells stimulated with PMA plus A23187 | |
| 0.1 mg | |
| Immunology | |
| Rat | |
| IgM κ |
| Flow Cytometry | |
| 1F4 | |
| PE | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.