Learn More
CD3 Mouse anti-Human, PE-CF594, Clone: UCHT1, BD
Mouse Monoclonal Antibody
£174.00 - £385.11
Specifications
| Antigen | CD3 |
|---|---|
| Clone | UCHT1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE-CF594 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15821809
|
BD Biosciences
562310 |
25 Tests |
£181.00 £174.00 / Each Save £7.00 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15821779
|
BD Biosciences
562280 |
100 Tests |
£401.00 £385.11 / Each Save £15.89 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The UCHT1 monoclonal antibody specifically binds to the human CD3ε-chain, a 20-kDa subunit of the CD3/T cell antigen receptor complex. CD3ε is expressed on 70-80% of normal human peripheral blood lymphocytes and 60-85% of thymocytes. Studies from the HLDA Workshop show that this antibody is mitogenic for CD3ε-positive cells when used in conjunction with costimulatory agents such as pokeweed mitogen or anti-CD28 antibody. CD3 plays a central role in signal transduction during antigen recognition. The UCHT1 antibody stains both surface and intracellular CD3ε unlike the other CD3 clone, HIT3a, that stains only extracellular CD3ε.
This antibody is conjugated to BD Horizon™ PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496nm and 564nm, PE-CF594 can be excited by the blue (488-nm), green (532-nm) and yellow-green (561-nm) lasers and can be detected with the same filter set as PE-Texas Red (eg 610/20-nm filter).
Specifications
| CD3 | |
| Flow Cytometry | |
| PE-CF594 | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified | |
| CD3e; CD3E; T3E; TCRE; T-cell surface antigen T3/Leu-4 epsilon |
| UCHT1 | |
| Monoclonal | |
| Mouse | |
| Human | |
| Human infant thymocytes and peripheral blood lymphocytes from a Sézary Syndrome donor | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.