missing translation for 'onlineSavingsMsg'
Learn More

CD3 Rat anti-Mouse, NA/LE, Unlabeled, Clone: 17A2, BD

Rat Monoclonal Antibody

Brand:  BD Biosciences 562163

Product Code. 15801699

  • ONLINE EXCLUSIVE PRICE
    £225.00
    £215.82 / Each
    Save £9.18
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The 17A2 monoclonal antibody specifically binds to the T-cell receptor-associated CD3 complex that is expressed on many thymocytes and mature T lymphocytes. Plate-bound 17A2 antibody has been reported to induce IL-2 production by cultured T cells in the absence of accessory cells. The binding of the 17A2 antibody to T cells can be blocked by the anti-CD3e mAb 145-2C11. This suggests that the 17A2 antibody recognizes an epitope of the CD3 epsilon chain. In vivo treatment with 17A2 antibody has been reported to partially deplete T lymphocytes and temporarily down-modulates CD3 expression on T cells.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD3
Monoclonal
1.0mg/mL
CD3; CD3 epsilon; Cd3e; CD3ε; T3e
Rat
Affinity Purified
RUO
Mouse
IgG2b κ
Flow Cytometry
17A2
Unconjugated
No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay.
Alphadelta TCR-positive T-T hybridoma D1
0.5 mg
Primary
Store undiluted at 4°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.