Learn More
CD3ε Hamster anti-Mouse, Biotin, Clone: 145-2C11, BD
Hamster Monoclonal Antibody
£32.53 - £322.97
Specifications
| Antigen | CD3 ε |
|---|---|
| Clone | 145-2C11 |
| Concentration | 0.5mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15826968
|
BD Biosciences
553059 |
0.1 mg |
£34.25 £32.53 / Each Save £1.72 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15836968
|
BD Biosciences
553060 |
0.5 mg |
£336.00 £322.97 / Each Save £13.03 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The 145-2C11 antibody reacts with the 25-kDa ε chain of the T-cell receptor-associated CD3 complex, which is expressed on thymocytes, mature T lymphocytes, and NK-T cells. The cytoplasmic domain of CD3e participates in the signal transduction events which activate several cellular biochemical pathways as a result of antigen recognition. Soluble 145-2C11 antibody can activate either unprimed (naive) or primed (memory/preactivated) T cells in vivo or in vitro, in the presence of Fc receptor-bearing accessory cells. In contrast, plate-bound 145-2C11 can activate T cells in the absence of accessory cells. Soluble 145-2C11 antibody has been reported to induce re-directed lysis of Fc receptor-bearing target cells by CTL clones and can also block lysis of specific target cells by antigen-specific CTL's. Under some conditions, T-cell activation by 145-2C11 antibody has been reported to result in apoptotic cell death. The 145-2C11 antibody does not cross-react with rat leukocytes and it has been reported that pre-incubation of thymus cell suspensions at 37°C for 2-4 hours prior to staining enhances the ability of anti-CD3ε and anti-αβ TCR mAbs to detect the T-cell receptor on immature thymocytes. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Hamster
Clone: 145-2C11
Isotype: IgG1 κ
Species Reactivity: Mouse
Immunogen: H-2Kb specific cytotoxic T lymphocyte clone BM10-37
Flow Cytometry
Specifications
| CD3 ε | |
| 0.5mg/mL | |
| Monoclonal | |
| Hamster | |
| Immunology | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified | |
| CD3; CD3 epsilon; Cd3e; CD3ε; T3e |
| 145-2C11 | |
| Flow Cytometry | |
| Biotin | |
| RUO | |
| Mouse | |
| H-2Kb specific cytotoxic T lymphocyte clone BM10-37 | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.