Learn More
CD3 Mouse anti-Human, AmCyan, Clone: SK7, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 339186
Description
CD3 clone SK7 is derived from hybridization of mouse NS-1 myeloma cells with spleen cells from BALB/c mice immunized with human thymocytes.
The CD3 antibody reacts with the epsilon chain of the CD3 antigen/T-cell antigen receptor (TCR) complex. This complex is composed of at least six proteins that range in molecular weight from 20 to 30 kilodaltons (kDa). The antigen recognized by CD3 antibodies is noncovalently associated with either α/β or γ/δ TCR (70 to 90 kDa).
Specifications
| CD3 | |
| Monoclonal | |
| 100.0μg/mL | |
| Phosphate buffered saline with BSA and 0.1% sodium azide. | |
| Human Thymocytes | |
| RUO | |
| Primary | |
| Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
| Flow Cytometry | |
| SK7 | |
| AmCyan | |
| Mouse | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.