Learn More
CD297 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542209
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: TLCYEVHYRT KDVHFNAYTG ATIRFGQFLS TSLLKEEAQE FGNQTLFTIF Sequence homology: Cow: 85%; Horse: 85%; Human: 100%; Pig: 85%; Rabbit: 92%; Rat: 79%; Sheep: 85%
CD297 Polyclonal antibody specifically detects CD297 in Human samples. It is validated for Western Blot.Specifications
CD297 | |
Polyclonal | |
Unconjugated | |
Art4 | |
4432404K01Rik; ADP-ribosyltransferase 4; ADP-ribosyltransferase 4 (DO blood group); ADP-ribosyltransferase 4 (Dombrock blood group); ADP-ribosyltransferase 4 (inactive) (Dombrock blood group); ADP-ribosyltransferase C2 and C3 toxin-like 4; ART4; ARTC4; ATR4; CD297; DO; DOK1; Dombrock blood group carrier molecule; ecto-ADP-ribosyltransferase 4; mono (ADP-ribosyl)transferase; mono(ADP-ribosyl)transferase 4; mono-ADP-ribosyltransferase 4; NAD(+)--protein-arginine ADP-ribosyltransferase 4; NAD(P)(+)--arginine ADP-ribosyltransferase; NAD(P)(+)--arginine ADP-ribosyltransferase 4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
420 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q93070 | |
Art4 | |
Synthetic peptide directed towards the middle region of human ART4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |