Learn More
CD29 Mouse anti-Human, PE, Clone: HUTS-21, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 556049
Description
Reacts with the 130 kDa integrin β 1 subunit, CD29, that is expressed following cell activation as a heterodimeric complex with one of nine distinct α subunits, comprising the very late activation antigen (VLA) subfamily of adhesion receptors. The HUTS-21 β 1 epitope is weakly expressed on resting lymphocytes, but is upregulated by TPA, Ca++ ionophore, or another CD29 antibody, TS2/16. This upregulation is Mn++ dependent and not supported by other divalent cations such as Ca++ and/or Mg++. The VLA receptors have a broad cellular distribution and interact with ligands like VCAm-1 and MAdCAM-1 during cell adhesion. They also interact with extra-cellular matrices including collagen, laminin, fibronectin and others. HUTS-21 antibody enhances adherence of TPA-activated lymphocytes to fibronectin.
Specifications
| CD29 | |
| Monoclonal | |
| PE | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| HUTS-21 | |
| Integrin β1 Chain | |
| Mouse | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.