missing translation for 'onlineSavingsMsg'
Learn More

CD29 Arminian Hamster anti-Mouse, Unlabeled, Clone: HM Tests1-1, BD

Product Code. 15837418
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15837418 0.5 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15837418 Supplier BD Biosciences Supplier No. 553837

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hamster Monoclonal Antibody

The HM β;1-1 monoclonal antibody specifically binds to the 130-kDa integrin β;1 chain (CD29). CD29 is expressed on the cell surface as a heterodimer with one of the distinct integrin-α; chains. With α;1 through α;6 (CD49a through CD49f), it forms the VLA-1 through VLA-6 complexes, respectively, and with α;v (CD51), it forms α;vβ;1 integrin. It also associates with the integrin α;7 α;8, and α;9 chains in non-lymphoid tissues. As a result, CD29 has a broad tissue distribution, including lymphocytes, endothelia, smooth muscle, epithelia, and oocytes. This hamster mAb to a mouse leukocyte antigen has been observed to crossreact with similar populations of rat leukocytes. Source of the immunogen was purified mouse VLA-4 (α;4β;1, CD49d/CD29).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD29
Applications Flow Cytometry
Classification Monoclonal
Clone HM Tests1-1
Concentration 0.5mg/mL
Conjugate Unconjugated
Description Itgb1; Integrin beta-1; Fnrb; gpIIa; VLA-4 subunit beta
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Hamster
Immunogen Purified mouse VLA-4
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.