missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA112824
This item is not returnable.
View return policy
Description
A suggested positive control for this product is placenta, spleen or peripheral blood leukocytes.
Toll-like receptors (TLRs) are signaling molecules that recognize different microbial products during infection and serve as an important link between the innate and adaptive immune responses (1-3). These proteins act through adaptor molecules such as MyD88 and TIRAP to activate various kinases and transcription factors such as Protein Kinase C (PKC) á/â and NF- êB (4-6). Studies with TLR4-deficient mice indicate that the main ligand for TLR is lipopolysaccharide. Consequently, these mice also showed increased susceptibility to Gram-negative sepsis.
Specifications
| TLR4 | |
| Polyclonal | |
| Unconjugated | |
| TLR4 | |
| Toll-like Receptor 4, ARMD10, CD284, TLR-4, TOLL, hToll, homolog of Drosophila toll | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7099 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunocytochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| O00206 | |
| TLR4 | |
| Synthetic peptide corresponding to 15 residues near the N-terminus of human TLR4. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction