missing translation for 'onlineSavingsMsg'
Learn More

anti-CD28 FITC, Clone: JJ319, Invitrogen™

CD28 Monoclonal antibody A16184 from Invitrogen specifically detects CD28 in Rat samples. It is validated for Flow Cytometry.

Brand:  Invitrogen A16184

Product Code. 15215944

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

CD28 Monoclonal antibody A16184 from Invitrogen specifically detects CD28 in Rat samples. It is validated for Flow Cytometry.

CD28 antigen is a 44 kDa disulfide linked homodimeric T cell specific surface glycoprotein. It is a cell adhesion molecule of the immunoglobulin superfamily which is constitutively expressed on most peripheral blood T lymphocytes (approximately 95% of CD4+ cells and 50% of CD8+ cells). Mature thymocytes exhibit higher levels of CD28 than the immature cells. Activation of T cells results in enhanced CD28 expression. T cell activation requires two combined signals provided by antigen presenting cells.
Host Species: Mouse
Cross Reactivity: Rat
Isotype: IgG
Clone: JJ319
Immunogen: Rat CD28
Formulation: 50μg Purified Antibody (0.5mg/ml) in proprietary buffer, with 0.09% sodium azide
Synonyms: MGC138290, Tp44, CD28 antigen, T-cell-specific surface glycoprotein CD28
Uniprot Accession: P31042
Entrez GeneID: 25660
Gene Symbol: Cd28
This product detects CD28 in Rat samples. This product has been successfully used in Flow Cytometry procedures. The immunogen is Rat CD28.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD28
Monoclonal
FITC
P31042
Cd28
Rat CD28
Primary
Rat
Purified
Flow Cytometry
JJ319
50μg Purified Antibody (0.5mg/ml) in proprietary buffer, with 0.09% sodium azide
MGC138290, Tp44, CD28 antigen, T-cell-specific surface glycoprotein CD28
Mouse
50 μg
25660
Store at 4° C
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.