missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD28 Hamster anti-Mouse, NA/LE, Unlabeled, Clone: 37.51, BD
Hamster Monoclonal Antibody
Brand: BD Biosciences 553294
This item is not returnable.
View return policy
Description
The 37.51 antibody reacts with CD28, which is expressed on most thymocytes, at low density on nearly all CD4+ and CD8+ peripheral T cells, and at even lower density on NK cells. The expression of CD28, in splenocytes and thymocytes,has been reported to increase after activation. CD28 transcripts are found in mast cells, and cell-surface expression of CD28 is induced upon maturation or activation of mast cells. It has been reported that CD28 is not expressed on some populations of intraepithelial T lymphocytes. CD28 is a costimulatory receptor; its ligands include CD80 (B7-1) and CD86 (B7-2). The 37.51 mAb augments proliferation and cytokine production by activated T and NK cells and can provide a costimulatory signal for CTL induction. There is considerable evidence that CD28 is a costimulatory receptor involved in many, but not all, T cell-dependent immune responses.
Specifications
| CD28 | |
| Monoclonal | |
| 1.0mg/mL | |
| Cd28; CD28 antigen; T-cell-specific surface glycoprotein CD28 | |
| Hamster | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
| Flow Cytometry | |
| 37.51 | |
| Unconjugated | |
| No azide/low endotoxin: Aqueous buffered solution containing protein stabilizer, no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay. | |
| Mouse EL-4 (T-cell lymphoma) Cells | |
| 0.5 mg | |
| Immunology | |
| Mouse | |
| IgG2 λ1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction