Learn More
CD273 Mouse anti-Human, PE, Clone: MIH18, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 558066
Description
The MIH18 monoclonal antibody specifically binds to CD273 which is also known as the Programmed Death Ligand 2 (PD-L2) and B7-DC. CD273 is a type 1 transmembrane glycoprotein member of the B7 family and Ig superfamily. CD273 serves as a ligand for CD279, the Programmed Death 1 (PD-1) receptor. CD273 is expressed by dendritic cells, activated monocytes, medullary thymic epithelial cells, placental trophoblasts, and myocardial endothelium. CD273 can function as a coinhibitor of T cell functions by acting through CD279. Monoclonal antibodies that block the CD273 and CD279 interaction result in enhanced T cell proliferation and cytokine production. Thus, CD273 can play an important role in regulating T cell responses. Clone MIH18 is reportedly a blocking antibody.
Specifications
| CD273 | |
| Monoclonal | |
| PE | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human CD273 Transfected Cell Line | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| MIH18 | |
| PD-1 ligand 2; PD-L2; B7-DC; PDCD1LG2; PDCD1 ligand 2; PDCD1L2; PDL2; Btdc | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.