Learn More
CD27 Mouse anti-Human, PE, Clone: M-T271, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 557330
Description
Clone M-T271 reacts with the human form of 110 kD disulfide-linked dimer of two polypeptide chains. CD27 molecule is a lymphocyte-specific member of the TNFR/NGFR family, and is expressed on the subset of human thymocytes and on the majority of mature T lymphocytes. This clone also cross-reacts with a major subset of peripheral blood lymphocytes and monocytes of baboon, and both rhesus and cynomolgus macaque monkeys. CD27+ cells were detected on all CD3+ lymphocytes and on a subset of CD20+ cells. The reactivity on lymphocytes and monocytes is similar to that observed with peripheral blood lymphocytes from normal human donors.
Specifications
| CD27 | |
| Monoclonal | |
| PE | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| M-T271 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human T-CLL cells | |
| 50 Tests | |
| Immunology | |
| Baboon, Cynomolgus, Rhesus Monkey | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.