Learn More
CD27 Mouse anti-Human, PE, Clone: L128, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 340425
Description
The CD27 antibody, clone L128, is derived from hybridization of mouse Sp2/0 cells with spleen cells from BALB/c mice immunized with activated peripheral blood lymphocytes.
The CD27 antibody recognizes a 110–120-kilodalton (kDa) disulfide-linked homodimer comprised of two 55-kDa polypeptide chains. The CD27 antigen is a lymphocyte-specific member of the tumor necrosis factor receptor (TNFR) super family, which also includes nerve growth factor receptor, CD30, CD40, CD95 (Fas), CD120a, CD120b, CD134 (OX 40), and CD137. The CD27 antigen is also known as S152, T14, TNFRSF7, and Tp55.
Specifications
| CD27 | |
| Monoclonal | |
| 1.6μg/mL | |
| Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
| Human Activated Peripheral Blood Cells | |
| RUO | |
| Primary | |
| Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
| Flow Cytometry | |
| L128 | |
| PE | |
| Mouse | |
| 50 Tests | |
| Immunology | |
| Human | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.