missing translation for 'onlineSavingsMsg'
Learn More

CD27 Mouse anti-Human, PE-CF594, Clone: M-T271, BD

Mouse Monoclonal Antibody

£180.38 - £415.28

Specifications

Antigen CD27
Clone M-T271
Applications Flow Cytometry
Classification Monoclonal
Conjugate PE-CF594
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15861819
View Documents
BD Biosciences
562324
25 Tests
ONLINE EXCLUSIVE PRICE
£188.00
£180.38 / Each
Save £7.62
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15891789
View Documents
BD Biosciences
562297
100 Tests
ONLINE EXCLUSIVE PRICE
£433.00
£415.28 / Each
Save £17.72
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The M-T271 monoclonal antibody specifically binds to CD27. CD27 presents asa type I transmembrane, disulphide-linked 110 kDa homodimer comprised of two polypeptide chains. The CD27 molecule is a lymphocyte-specific member of the TNF/NGF-R family, and is expressed on a subset of human thymocytes and on the majority of mature T lymphocytes, activated B cells and NK cells. CD27 is highly induced on T cells after TCR stimulation. CD27 binds to CD70 (also known as, CD27 ligand or CD27L) and may be involved in cellular interaction of T and B lymphocytes.

This antibody is conjugated to BD Horizon™ PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496nm and 564nm, PE-CF594 can be excited by the blue (488-nm), green (532-nm) and yellow-green (561-nm) lasers and can be detected with the same filter set as PE-Texas Red (eg 610/20-nm filter).

Specifications

Specifications

CD27
Flow Cytometry
PE-CF594
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
IgG1 κ
Affinity Purified
TNFRSF7; TNF receptor superfamily, member 7; T14; Tp55; S152
M-T271
Monoclonal
Mouse
Human
Human T-CLL cells
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.