Learn More
anti-CD27/TNFRSF7 Clone: 137B4
CD27/TNFRSF7 Monoclonal Ab MA5-12191 from Thermo Scientific™ specifically detects CD27/TNFRSF7 in Hu samples, and it is validated for Flow Cytometry, IHC(P) and WB
Brand: Thermo Scientific MA5-12191
Description
CD27/TNFRSF7 Monoclonal Ab MA5-12191 from Thermo Scientific™ specifically detects CD27/TNFRSF7 in Hu samples, and it is validated for Flow Cytometry, IHC(P) and WB
CD27 (also known as S152 or T14).is expressed on the majority of peripheral T cells, medullary thymocytes, memory-type B cells, and natural killer cells. It is a transmembrane phosphoglycoprotein that belongs to the tumor necrosis factor receptor (TNFR) superfamily. CD27 binds to its ligand CD70, a member of the TNF family, and induces T-cell costimulation and B-cell activation. It also interacts with TRAFs and is involved in activation of NFdeltaB and SAPK/JNK and induces apoptosis.Host Species: Mouse
Cross Reactivity: Human
Isotype: IgG1
Clone: 137B4
Immunogen: Recombinant human CD27 protein
Formulation: 500μL unpurified Antibody in tissue culture supernatant, with 0.09% sodium azide;
Synonyms: CD27 antigen, CD27L receptor, T cell activation antigen CD27, T cell activation antigen S152, T-cell activation antigen CD27, tumor necrosis factor receptor superfamily member 7
Uniprot Accession: P26842
Gene Symbol: CD27
Entrez GeneID: 939
MA5-12191 detects CD27/TNFRSF7 in Human samples. MA5-12191 has been successfully used in Flow Cytometry, Immunohistochemistry (Paraffin) and Western Blot procedures. MA5-12191 The immunogen is Recombinant human CD27 protein.
Specifications
| CD27/TNFRSF7 | |
| Monoclonal | |
| Unconjugated | |
| P26842 | |
| CD27 | |
| Recombinant human CD27 protein | |
| RUO | |
| 939 | |
| Store at 4° C | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 137B4 | |
| 500μL unpurified Antibody in tissue culture supernatant, with 0.09% sodium azide; | |
| CD27 antigen, CD27L receptor, T cell activation antigen CD27, T cell activation antigen S152, T-cell activation antigen CD27, tumor necrosis factor receptor superfamily member 7 | |
| Mouse | |
| 500 μL | |
| Primary | |
| Human | |
| Supernatant |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.