missing translation for 'onlineSavingsMsg'
Learn More

CD27 Mouse anti-Human, APC, Clone: M-T271, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 561400

Product Code. 15861009

  • ONLINE EXCLUSIVE PRICE
    £199.00
    £196.88 / Each
    Save £2.12
    1% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The M-T271 monoclonal antibody specifically binds to the human form of CD27, a type I transmembrane, 110 kDa disulfide-linked homodimeric glycoprotein. The CD27 molecule is a lymphocyte-specific member of the TNFR/NGFR family. CD27 is expressed on a subset of human thymocytes and on the majority of mature T lymphocytes. This antibody also crossreacts with a major subset of peripheral blood lymphocytes and monocytes obtained from some non-human primates (NHP) including baboons as well as rhesus and cynomolgus macaques. CD27 expression is detected on the majority of CD3+ lymphocytes and on a subset of CD20+ cells derived from these NHP donors. The CD27 staining patterns on lymphocytes and monocytes from these NHP donors is similar to that observed with peripheral blood lymphocytes from normal human donors.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD27
Monoclonal
APC
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human T-CLL cells
50 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
M-T271
TNFRSF7; TNF receptor superfamily, member 7; T14; Tp55; S152
Mouse
Affinity Purified
RUO
Baboon, Cynomolgus, Rhesus Monkey
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.