Learn More
CD258 (LIGHT) Mouse anti-Human, Alexa Fluor 647, Clone: 115520, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 564374
Description
The 115520 monoclonal antibody specifically binds to CD258 which is also known as LIGHT (is homologous to lymphotoxins, exhibits inducible expression, and competes with HSV glycoprotein D for HVEM, a receptor expressed by T lymphocytes). CD258 is also referred to as HVEM-L (Herpes virus entry mediator ligand), and LT- γ. This type II transmembrane glycoprotein belongs to TNF superfamily; it is encoded by TNFSF14 (tumor necrosis factor ligand superfamily member 14). CD258 is expressed by activated T cells and cultured dendritic cells. CD258 can costimulate T cells and antigen presenting cells as well as trigger apoptosis by some cell types. CD258 binds to CD270/LIGHT-R/HVEM and to the Lymphotoxin β Receptor (LT β R). LIGHT reportedly also binds to the decoy receptor (DcR3/TR6).
Specifications
| CD258 (LIGHT) | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human LIGHT Recombinant Protein | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 115520 | |
| TNFSF14; LIGHT; HVEML; LTg | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.