missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD244 Monoclonal Antibody (2B4.69), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA516486
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This product is a low-endotoxin, preservative-free formulation. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
CD244 Monoclonal antibody specifically detects CD244 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation.Specifications
CD244 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q9BZW8 | |
CD244 | |
CD244 Fusion protein. | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunoprecipitation | |
2B4.69 | |
Unconjugated | |
CD244 | |
2B4; C9.1; CD244; CD244 long isoform; CD244 molecule; Cd244 molecule, natural killer cell receptor 2B4; CD244 natural killer cell receptor 2B4; CD244 short isoform; F730046O15Rik; h2B4; Ly90; NAIL; natural killer cell receptor 2B4; NK cell activation inducing ligand NAIL; NK cell activation-inducing ligand; NK cell receptor 2B4; NK cell type I receptor protein 2B4; NKR Nmrk; NKR2B4; Nmrk; non MHC restricted killing associated; non-MHC restricted killing associated; Signaling lymphocytic activation molecule 4; SLAM family member 4; SLAMF4; transmembrane NK cell receptor 2B4 | |
Mouse | |
Protein G | |
RUO | |
51744 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |