Learn More
CD226 Mouse anti-Human, NA/LE, Unlabeled, Clone: DX11, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 559786
Description
Reacts with DNAX accessory molecule-1 (DNAM-1), a 65kDa transmembrane glycoprotein consisting of 318 amino acid residues including two Ig-like domains. DNAM-1 is expressed on the majority of T cells, NK cells, monocytes, and a subset of B cells, but not on granulocytes or erythrocytes. It is also present on a subset of thymocytes coexpressing high density surface CD3. DNAM-1 is not present on normal fibroblast cell lines or tumor cell lines of epithelial or neuronal origins. DNAM-1 is a tyrosinephosphorylated, signal-transducing molecule which participates in primary adhesion during cytotoxic T lymphocyte (CTL)-mediated cytotoxicity. DX11 inhibits T- and NKcell-mediated cytotoxicity against a variety of tumor cell targets, and blocks cytokine production by alloantigen-specific T cells.
Flow Cytometry
Specifications
| CD226 | |
| Monoclonal | |
| 1.0mg/mL | |
| DNAM-1; DNAM1; PTA1; PTA-1; TLiSA1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
| Flow Cytometry | |
| DX11 | |
| Unconjugated | |
| No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay. | |
| Human CTL Clone | |
| 0.5 mg | |
| Immunology | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.