Learn More
CD22.2 Mouse anti-Mouse, Biotin, Clone: CY34.1, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 553382
Description
The Cy34.1 antibody reacts with the B-lymphocyte differentiation antigen CD22 on strains having the Lyb-8.2 alloantigen (e.g., A, BALB/c, CBA, C3H/He, C57BL, C57L, C58, SJL, SWR, but not AKR, DBA/1, DBA/2, NZB, PL). CD22 is expressed at high levels on mature peripheral B lymphocytes (follicularand marginal zone), B-1 cells (CD5+ B cells), and plasma cells. It is a member of the Ig gene superfamily and associates with the B-cell antigen receptor. Its sialic acid- binding immunoglobulin-like lectin (siglec) extracellular region mediates B-cell adhesion to ligands on endothelial cells in the bone marrow. Its intracellulardomain is phosphorylated after cross-linking of antigen receptoror MHC class II antigen. It is involved in negative regulation of B-cell activation and protection from autoimmunity. B-cell proliferative responses to LPS or anti-mouse Ig µ chain are augmented in the presence of Cy34.1 mAb.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in literature.
Specifications
| CD22.2 | |
| Monoclonal | |
| 0.5mg/mL | |
| Lyb-8.2 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| CY34.1 | |
| Biotin | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| B10.D2 mouse splenocytes | |
| 0.5 mg | |
| Immunology | |
| Mouse | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.