Learn More
CD212 Mouse anti-Mouse, Biotin, Clone: 114, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 551973
Description
The 114 antibody reacts with mouse CD212 (the β1 subunit of IL-12Rβ1), originally termed IL-12Rβ, of the mouse IL-12 receptor complex. The IL-12Rβ1 subunit associates with a β2 subunit to form a heterodimeric IL-12 receptor complex. Each one of the IL-12R subunits exhibits low affinity for IL-12, but in combination, they bind IL-12 with high affinity. The IL-12Rβ1 subunit interacts primarily with IL-12 p40 whereas the IL-12Rββ1 has more recently been described to bind IL-23, a heterodimer formed of the p40 subunit from IL-12, and p19. The cytoplasmic regions of the β1 and β2 subunits contain the box1 and box2 motifs found in other cytokine receptors such as gp130, LIFR and G-CSFR. IL-12Rβ1 are primarily expressed by activated T cells and NK cells. Experiments with IL-12Rβ1 deficient mice have shown that IL-12Rβ1 is necessary for mouse T and NK cell responsiveness to IL-12 p75. The 114 antibody was generated by immunizing IL-12Rβ1 deficient mice of (129 x BALB/c)F1 background with mouse Ba/F3 cells that were stably transfected with IL-12Rβ1
Specifications
| CD212 | |
| Monoclonal | |
| 0.5mg/mL | |
| IL-12 Receptor β1 chain | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 114 | |
| Biotin | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IL-12Rβ1 Transfected Cell Line | |
| 0.1 mg | |
| Immunology | |
| Mouse | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.