missing translation for 'onlineSavingsMsg'
Learn More

CD206 Mouse anti-Human, Unlabeled, Clone: 19.2, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 555953

Product Code. 15878548

  • ONLINE EXCLUSIVE PRICE
    £125.00
    £119.58 / Each
    Save £5.42
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reacts with the macrophage mannose receptor (MR), a membrane glycoprotein of approximately 162kDa which binds to glycoconjugates containing mannose, fucose, or N-acetylglucosamine. These carbohydrates are present on the surface of many microorganisms and enable the macrophage to bind to microorganisms through the MR and internalize them during the process of phagocytosis. Mannose receptors are expressed on human macrophages and cultured dendric cells. They are not detected on resting moncytes. Mannose receptor expression is upregulated on PBMC following 3 day incubation with GM-CSF. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD206
Monoclonal
0.5mg/mL
Macrophage mannose receptor
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C
Flow Cytometry
19.2
Unconjugated
Aqueous buffered solution containing ≤0.09% sodium azide.
Human Placental Macrophage Mannose Receptor
0.1 mg
Immunology
Human
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.