Learn More
CD205 Mouse anti-Human, Unlabeled, Clone: MMRI-7, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 557778
Description
Reacts with DEC-205, a 205 kDa transmembrane protein with C-type lectin external domains, present on dendritic cells (DC) and some epithelia. It is being described as a multilectin receptor for adsorptive endocytosis. Reports indicate that CD205 (DEC-205) mRNA levels increase significantly during activation. In monocyte derived dendritic cells, the expression of CD205 increased during maturation. Antibody MMRI-7 is useful for the study of DC development.
Specifications
| CD205 | |
| Monoclonal | |
| 0.5mg/mL | |
| DEC-205 | |
| Mouse | |
| 0.1 mg | |
| Immunology | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| MMRI-7 | |
| Unconjugated | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.