Learn More
CD205 Mouse anti-Human, Unlabeled, Clone: MG38, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 558531
Description
Reacts with DEC-205, a 205 kDa transmembrane protein with C-type lectin external domains, present on dendritic cells (DC) and some epithelia. It is being described as a multilectin receptor for adsorptive endocytosis. MG38 antibody reacted with mature monocyte-derived DC and also with B cells, monocytes and weakly on T cells and NK cells. Antibody MG38 is useful for the study of DC development.
Specifications
| CD205 | |
| Monoclonal | |
| 0.5mg/mL | |
| DEC-205; Lymphocyte Antigen 75; CLEC13B | |
| Mouse | |
| 0.1 mg | |
| Immunology | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| MG38 | |
| Unconjugated | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.