Learn More
CD204 Monoclonal Antibody (2F8), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA516492
Code : NEW
Additional Details : Weight : 0.01000kg
Description
In Western blot, this product recognizes CD204 in J774 cells under non-reducing conditions only. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
CD204 Monoclonal antibody specifically detects CD204 in Fish, Mouse, Porcine samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot.Specifications
CD204 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P30204 | |
MSR1 | |
Raw 264 cell line | |
250 μg | |
Primary | |
Fish, Mouse, Porcine | |
Antibody | |
IgG2b |
ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
2F8 | |
Unconjugated | |
MSR1 | |
bovine scavenger receptor type II; CD204; Macrophage acetylated LDL receptor I and II; macrophage scavenger receptor 1; macrophage scavenger receptor type III; macrophage scavenger receptor types I and II; MRS-A; MSR; Msr1; MSR-A; phSR1; phSR2; RGD1564316; SCARA1; Scavenger receptor class A member 1; scavenger receptor class A, member 1; scavenger receptor type A; scavenger receptor type I; Scvr; SRA; SR-A; SR-AI; SR-AI/II; SR-AII | |
Rat | |
Protein G | |
RUO | |
100511665, 20288 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |