missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD203c Monoclonal Antibody (NP4D6), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA110138
Code : LD
Additional Details : Weight : 0.01000kg
Description
The protein encoded by this gene belongs to a series of ectoenzymes that are involved in hydrolysis of extracellular nucleotides. These ectoenzymes possess ATPase and ATP pyrophosphatase activities and are type II transmembrane proteins. Expression of the related rat mRNA has been found in a subset of immature glial cells and in the alimentary tract. The corresponding rat protein has been detected in the pancreas, small intestine, colon, and liver. The human mRNA is expressed in glioma cells, prostate, and uterus. Expression of the human protein has been detected in uterus, basophils, and mast cells. [provided by RefSeq, Jul 2008]Specifications
CD203c | |
Monoclonal | |
1.0 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
O14638 | |
ENPP3 | |
HEK-293 cells transfected with human CD203c | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry | |
NP4D6 | |
Unconjugated | |
ENPP3 | |
AI876438; alkaline phosphodiesterase; Alkaline phosphodiesterase I; B10; CD203c; dJ1005H11.3 (phosphodiesterase I/nucleotide pyrophosphatase 3); dJ914N13.3 (phosphodiesterase I/nucleotide pyrophosphatase 3); ectonucleotide pyrophosphatase/phosphodiesterase 3; ectonucleotide pyrophosphatase/phosphodiesterase family member 3; E-NPP 3; ENPP3; gp130RB13-6; Npp3; NPPase; Nucleotide diphosphatase; Nucleotide pyrophosphatase; PD-Ibeta; Pdnp3; Phosphodiesterase I beta; phosphodiesterase I/nucleotide pyrophosphatase 3; phosphodiesterase-I beta; RB13-6; RB13-6 antigen | |
Mouse | |
Protein A | |
RUO | |
5169 | |
4° C, do not freeze | |
Liquid |