Learn More
CD203c Mouse anti-Human, Brilliant Violet 421, Clone: NP4D6, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 563296
Description
The NP4D6 monoclonal antibody specifically binds to CD203c. CD203c is a type II transmembrane glycoprotein that is a member of the E-NNP family of ectoenzymes. CD203c is also known as ectonucleotide pyrophosphatase/phosphodiesterase 3 (E-NPP3, ENPP3) due to its capacity to hydrolyze phosphodiester and phosphosulfate bonds in a variety of molecules including deoxynucleotides, nucleoside phosphates, and nicotinamide adenine dinucleotide. CD203c is otherwise known as Phosphodiesterase-1 β (PD-1 β , PD-I beta), B10, or gp130RB13-6. CD203c is expressed by basophils and mast cells. Basophils increase CD203c expression following activation with allergens or antibodies that crosslink cytophilic IgE. Thus, CD203c serves as a useful flow cytometric marker for basophil activation and the detection and analysis of type 1 allergic responses. IgE-receptor cross-linking results in CD203c upregulation and overexpression on neoplastic mast cells in cases of systemic mastocytosis.
Specifications
| CD203c | |
| Monoclonal | |
| Brilliant Violet 421 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human CD203c Transfected Cell Line | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| NP4D6 | |
| ENPP3; E-NPP 3; NPP3; PDNP3; PD-Ibeta; PD-1β; B10; NPPase; gp130RB13-6 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.