Learn More
CD20 Mouse anti-Human, FITC, Clone: 2H7, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 556632
Description
Clone 2H7 reacts with the human form of a phosphoprotein, CD20, found on circulating peripheral blood B lymphocytes. This clone also cross-reacts with a subset pf peripheral blood lymphocytes, but not monocytes nor granulocytes, of baboon and both rhesus and cynomolgus macaque monkeys. The distrubution on lymphocytes is similar to that seen with normal human donor lyphocyes, namely bright staining on B lymphocytes and weak reactivity on a small subset of CD3-positive T lymphocytes.
Specifications
| CD20 | |
| Monoclonal | |
| FITC | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry, Immunohistochemistry (Frozen) | |
| 2H7 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human 6.16c1.3 B cell line | |
| 50 Tests | |
| Immunology | |
| Baboon, Cynomolgus, Rhesus Monkey | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.