Learn More
CD20 Mouse anti-Human, Unlabeled, Clone: H1, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555677
Description
The H1 (FB1) antibody specificially binds to a cytoplasmic domain of CD20. CD20 is a 33-37-kDa four transmembrane phosphoprotein that is expressed by B lymphocytes from the pre-B stage and most malignant B cells and is lost during plasma cell differentiation. Low level CD20 expression is observed on a subset of normal circulating T lymphocytes, and CD20-positive T-cell lymphomas have been reported. The CD20 molecule is associated with membrane lipid raft domains, acts as a channel for calcium ions, and is involved in the regulation of B cell activation and survival. The cytoplasmic domain regions are serine and threonine rich and contain multiple phosphorylation consensus sequences.
Specifications
| CD20 | |
| Monoclonal | |
| 0.5mg/mL | |
| MS4A1; membrane-spanning 4-domains subfamily A member 1; B1; Bp35; LEU-16 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
| Flow Cytometry | |
| H1 | |
| Unconjugated | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human B lymphoma cell line | |
| 0.1 mg | |
| Immunology | |
| Human | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.