missing translation for 'onlineSavingsMsg'
Learn More

CD1d Mouse anti-Human, Unlabeled, Clone: CD1D42, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 550254

Product Code. 15816198

  • ONLINE EXCLUSIVE PRICE
    £133.00
    £127.90 / Each
    Save £5.10
    3% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The CD1d42 monoclonal antibody recognizes CD1d. Cell surface CD1d is structurally homologous to Class I MHC molecules. It consists of a glycosylated type I transmembrane α chain (43-49 kDa) that is non-covalently associated with β2-microglobulin. CD1d is a member of the CD1 family of molecules, which belong to the immunoglobulin superfamily. Sequence homology data classifies the CD1 molecules into two groups. Group 1 includes CD1a, CD1b and CD1c molecules; group 2 includes CD1d molecules and their homologs in other species. CD1d is expressed on cortical thymocytes, B cells, dendritic cells, monocytes, and some nonlymphoid cells including intestinal epithelial cells, hepatocytes and keratinocytes. It is not expressed on resting mature T cells. Studies suggest that CD1d participates in lipid antigen presentation to CD1d-restricted NKT cells.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD1d
Monoclonal
0.5mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Human CD1d Recombinant Protein
0.1 mg
Immunology
Human
IgG1 κ
Flow Cytometry
CD1D42
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.