missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Monoclonal Antibody (LT19), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119242
Code : LD
Additional Details : Weight : 0.01000kg
Description
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow Cytometry, Immunoprecipitation.Specifications
CD19 | |
Monoclonal | |
1.0 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P15391 | |
Cd19 | |
B lymphoblast cell line Daudi. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunoprecipitation | |
LT19 | |
Unconjugated | |
Cd19 | |
AW495831; B4; B-lymphocyte antigen CD19; B-lymphocyte surface antigen B4; Cd19; CD19 antigen; CD19 molecule; CVID3; differentiation antigen CD19; Leu-12; T-cell surface antigen Leu-12 | |
Mouse | |
Protein A | |
RUO | |
930 | |
4° C, do not freeze | |
Liquid |