Learn More
CD19 Mouse anti-Human, APC-H7, Clone: SJ25C1, BD
Mouse Monoclonal Antibody
£161.79 - £358.08
Specifications
| Antigen | CD19 |
|---|---|
| Clone | SJ25C1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | APC-H7 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15880049
|
BD Biosciences
560252 |
25 Tests |
£168.00 £161.79 / Each Save £6.21 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15859988
|
BD Biosciences
560177 |
100 Tests |
£363.00 £358.08 / Each Save £4.92 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SJ25C1 antibody reacts with CD19, a B lymphocyte-lineage differentiation antigen. CD19, a 90kDa transmembrance glycoprotein, is a member of the immunoglobulin superfamily and is expressed throughout B-lymphocyte development from the pro-B cell through the mature B-cell stages. Terminally differentiated plasma cells do not express CD19. On the surface of mature B cells, the CD19 molecule associates with CD21 (CR-2) and CD81 (TAPA-1), and this multimolecular complex synergizes with surface immunoglobulin to promote cellular activation. Studies with CD19-deficient mice have suggested that the level of CD19 expression affects the generation and maturation of B cells in the bone marrow and periphery. B-1 lineage B cells, also known as CD5+ B cells, are drastically reduced or absent in CD19-deficient mice. Increased levels of CD19 expression correlate with increased frequencies of peritonal and splenic B-1 cells and reduced numbers of conventional B lymphocytes in the periphery. CD19 participates in B-lymphocyte development, B-cell activation, maturation of memory B cells and regulation of tolerance. CD19 has also been detected on peritoneal mast cells, co-localized with CD21/CD35, and it is proposed to play a role in complement-mediated mast-cell activation.
Host Species: Mouse
Clone: SJ25C1
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Specifications
| CD19 | |
| Flow Cytometry | |
| APC-H7 | |
| RUO | |
| Human | |
| Human NALM1 + NALM16 cells | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| SJ25C1 | |
| Monoclonal | |
| Mouse | |
| Immunology | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.