Learn More
CD19 Rat anti-Mouse, APC-H7, Clone: 1D3, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 560245
Description
The 1D3 antibody reacts with CD19, a B lymphocyte-lineage differentiation antigen. CD19, a 95kDa transmembrance glycoprotein, is a member of the immunoglobulin superfamily and is expressed throughout B-lymphocyte development from the pro-B cell through the mature B-cell stages. Terminally differentiated plasma cells do not express CD19. On the surface of mature B cells, the CD19 molecule associates with CD21 (CR-2) and CD81 (TAPA-1), and this multimolecular complex synergizes with surface immunoglobulin to promote cellular activation. Studies with CD19-deficient mice have suggested that the level of CD19 expression affects the generation and maturation of B cells in the bone marrow and periphery. B-1 lineage B cells, also known as CD5+ B cells, are drastically reduced or absent in CD19-deficient mice. Increased levels of CD19 expression correlate with increased frequencies of peritonal and splenic B-1 cells and reduced numbers of conventional B lymphocytes in the periphery. CD19 participates in B-lymphocyte development, B-cell activation, maturation of memory B cells and regulation of tolerance. CD19 has also been detected on peritoneal mast cells, co-localized with CD21/CD35, and it is proposed to
play a role in complement-mediated mast-cell activation.
Host Species: Rat
Clone: 1D3
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: Mouse CD19 Transfected Cell Line
Flow Cytometry, Intracellular Staining
Specifications
| CD19 | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
| Mouse CD19 Transfected Cell Line | |
| 25 μg | |
| Immunology | |
| Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| 1D3 | |
| APC-H7 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.