Learn More
CD182 Mouse anti-Human, PE, Clone: 6C6, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555933
Description
Reacts with interleukin-8 receptor B (IL-8RB), a G-protein-linked seven transmembrane domain protein, expressed on monocytes and granulocytes. It is also expressed on a subset of T cells. Both IL-8RA (CXCR1) and IL-8RB (CXCR2) are high affinity receptors for IL-8. IL-8 is an inflammatory cytokine which induces chemotaxis of neutrophils, basophils and T lymphocytes.
Specifications
| CD182 | |
| Monoclonal | |
| 0.2mg/mL | |
| CXCR2; CXCR-2; IL8RB; IL-8Rβ; Interleukin 8 receptor, beta; CDw128b | |
| Mouse | |
| 0.2 mg | |
| Immunology | |
| Human | |
| IgG1 λ |
| Flow Cytometry | |
| 6C6 | |
| PE | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.