Learn More
CD180 Mouse anti-Human, PE, Clone: G28-8, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 551953
Description
Reacts with RP105/Bgp95, a 95-105 kDa type I membrane protein consisting of extracellular leucine-rich repeats and a short cytoplasmic domain. It is expressed on mantle zone B cells, but weakly on germinal center B cells. RP105/Bgp95 is also expressed on peripheral blood monocytes, dendritic cells, and a subset of peripheral blood lymphocytes. The extracellular domain associates with a molecule called MD-1 to form a cell surface receptor complex RP105/Bgp95/MD- 1. This receptor belongs to the family of toll-like receptors (TLR). Studies show that RP105/Bgp95/MD-1, working in concert with TLR4, controls B cell recognition and signaling of lipopolysaccharide (LPS). Reports on functional studies show that G28-8 monoclonal antibody can induce a G0 to G1 cell cycle transition and was synergistic with PMA, anti-µ, or anti-CD40 in inducing proliferation of resting B cells.
Specifications
| CD180 | |
| Monoclonal | |
| PE | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| G28-8 | |
| RP105 | |
| Mouse | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.