Learn More
CD18 Mouse anti-Human, Unlabeled, Clone: L130, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 556084
Description
The L130 monoclonal antibody specifically binds to β 2 integrin, a 90-95 kDa type-1 transmembrane protein, which associates with the α chains of CD11a, CD11b, or CD11c as a heterodimer. CD18, known as lymphocyte function-associated antigen-1 (LFA-1), is expressed on lymphocytes, monocytes, and more weakly, on granulocytes. LFA-1 has been shown to play an important role in homotypic and heterotypic cellular adhesion in immune and inflammatory responses
Specifications
| CD18 | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
| Flow Cytometry | |
| L130 | |
| Unconjugated | |
| Mouse | |
| 0.1 mg | |
| Immunology | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.