Learn More
CD166 Mouse anti-Human, Unlabeled, Clone: 3A6, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 559260
Description
The 3A6 monoclonal antibody specifically binds to the activated leukocyte cell adhesion molecule (ALCAM), a 100-105 kDa transmembrane glycoprotein, also known as the CD6 ligand, CD166. It is composed of an immunoglobulin-like extracellular domain, a transmembrane region and a short cytoplasmic tail. ALCAM belongs to the Ig superfamily of proteins and is expressed on neurons, activated T cells, activated monocytes, epithelial cells and fibroblasts. CD166 plays an important role in mediating adhesion interactions between thymic epithelial cells and CD6+ cells during intrathymic T-cell development.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
| CD166 | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
| Flow Cytometry | |
| 3A6 | |
| Unconjugated | |
| Mouse | |
| 0.1 mg | |
| Immunology | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.