Learn More
CD164 Mouse anti-Human, Unlabeled, Clone: N6B6, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 551296
Description
Reacts with an 80-90 kDa, mucin-like molecule present on CD34+ cells during the early stages of B cell, erythroid cell and myelomonocytic cell development. It is also expressed on epithelial cells, peripheral blood monocytes and weakly on lymphocytes. Mucin-like molecules are a new family of glycoproteins present in tissues of the hematopoietic system. They are highly glycosylated polypeptides, containing predominantly O-linked carbohydrate side chains. Reports suggest that CD164 may play a role in hematopoiesis by facilitating the adhesion of CD34+ cells to bone marrow stroma.
This antibody is routinely tested by flow cytrometric analysis. Other applications were tested at BD Bioscience Pharmingen during antibody development only or reported in the literature.
Specifications
| CD164 | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human Pre-B Cell Line | |
| 0.1 mg | |
| Immunology | |
| Human | |
| IgG2a κ |
| Flow Cytometry | |
| N6B6 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.