missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD163 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA514215
Code : LE
Additional Details : Weight : 0.01000kg
Description
CD163 Polyclonal antibody specifically detects CD163 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
CD163 | |
Polyclonal | |
Unconjugated | |
CD163 | |
CD163; CD163 antigen; CD163 molecule; CD163v2; CD163v3; Hemoglobin scavenger receptor; M130; macrophage-associated antigen; MM130; putative CD163 antigen; SCARI1; Scavenger receptor cysteine-rich type 1 protein M130; sCD163; Soluble CD163; Soluble sCD163 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
9332 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
Q86VB7 | |
CD163 | |
KLH conjugated synthetic peptide between 245-274 amino acids from the N-terminal region of human CD163 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |