Learn More
CD162 Rat anti-Mouse, PE, Clone: 2PH1, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 555306
Description
The 2PH1 monoclonal antibody specifically binds to the N-terminus of CD162 (P-selectin glycoprotein ligand-1, PSGL-1), encoded by the Selplg gene. PSGL-1 is expressed on the cell surface as a homodimer of approximately 230 kDa. In the mouse, Selpl mRNA is detected in most tissues, with high levels found in hematopoietic cells, brain, and adipose tissue. Flow cytometric analyses have revealed CD162 expression on bone marrow-derived mast and dendritic cells, splenic leukocytes, platelets, peripheral blood neutrophils, and neutrophil and T-cell lines. PSGL-1 is a ligand for P-selectin (CD62P) and is involved in leukocyte rolling, the migration of leukocytes into inflamed tissues, and responses to vascular injury. It is a sialomucin that must be specifically sialylated, fucosylated, and sulfated to bind P-selectin. There is also evidence that other ligands for PSGL-1 and CD62P may exist. The 2PH1 antibody is reported to block binding of mouse leukocytes to CD62P, but the 4RA10 antibody (Cat. No. 557787) has significantly greater blocking activity.
Specifications
| CD162 | |
| Monoclonal | |
| 0.2mg/mL | |
| Selplg; PSGL-1; Psgl1; Selp1; Selpl; P-selectin glycoprotein ligand 1 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 2PH1 | |
| PE | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Ovalbumin-conjugated peptide covering amino acids 42 to 60 of mouse PSGL-1 | |
| 0.2 mg | |
| Immunology | |
| Mouse | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.