Learn More
CD160 Rat anti-Mouse, PE-CF594, Clone: CNX46-3, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 562512
Description
The CNX46-3 monoclonal antibody specifically binds to mouse CD160, also known as Natural Killer Cell Receptor BY55. CD160 is a glycosyolphosphatidylinositol-anchored membrane glycoprotein that contains a single Ig domain. This member of the Ig Superfamily was the first identified as a MHC Class I antigen-specific Ig-like Receptor that was expressed by mouse NK cells. As a receptor, CD160 can bind to classical and non-classical MHC class I molecules with low affinity. As a ligand, CD160 can bind to HVEM (Herpes Virus Entry Mediator, a TNF Receptor Family Member). CD160 is expressed on a subset of NK cells, NKT cells, activated CD8+ T cells and TCR γδ T cells. The functions of CD160 have been reported to regulate NK cell activation both positively and negatively, depending on stimulus.
This antibody is conjugated to BD Horizon™ PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red™. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red™ yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496nm and 564nm, PE-CF594 can be excited by the blue (488-nm), green (532-nm) and yellow-green (561-nm) lasers and can be detected with the same filter set as PE-Texas Red™ (eg 610/20-nm filter).
Specifications
| CD160 | |
| Monoclonal | |
| 0.2mg/mL | |
| Cd160; By55; Natural killer cell receptor BY55 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| CNX46-3 | |
| PE-CF594 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse CD160 Recombinant Protein | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.