missing translation for 'onlineSavingsMsg'
Learn More

CD16 Mouse anti-Human, PE-Cy7, Clone: 3G8, BD

Mouse Monoclonal Antibody

£56.71 - £273.92

Specifications

Antigen CD16
Clone 3G8
Applications Flow Cytometry
Classification Monoclonal
Conjugate PE-Cyanine7
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15820599
View Documents
BD Biosciences
560918
25 Tests
ONLINE EXCLUSIVE PRICE
£78.15
£74.11 / Each
Save £4.04
5% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15870429
View Documents
BD Biosciences
560716
50 Tests
ONLINE EXCLUSIVE PRICE
£57.55
£56.71 / Each
Save £0.84
1% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15839028
View Documents
BD Biosciences
557744
100 Tests
ONLINE EXCLUSIVE PRICE
£285.00
£273.92 / Each
Save £11.08
3% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Reacts with the 50-65kDa transmembrane form of IgG Fc (FcγRIII), a human NK-cell-associated antigen. CD16 is expressed on NK cells as well as macrophages and granulocytes. Reports indicate that CD16 plays a role in signal transduction and NK cell activation. CD16 antibody blocks the binding of soluble immune complexes to granulocytes. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: 3G8
Isotype: IgG1 κ
Species Reactivity: Human

Flow Cytometry

Specifications

Specifications

CD16
Flow Cytometry
PE-Cyanine7
RUO
Baboon, Cynomolgus, Human, Rhesus Monkey
Human polymorphonuclear leukocytes
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
3G8
Monoclonal
Mouse
Immunology
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
IgG1 κ
Affinity Purified
FcRIII; Fc-gamma RIII; FCG3; FCGR3; FCGRIII; FcγRIII; IGFR3
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.