missing translation for 'onlineSavingsMsg'
Learn More

CD16/CD32 Rat anti-Mouse, FITC, Clone: 2.4G2, BD

Rat Monoclonal Antibody

£151.60 - £417.13

Specifications

Antigen CD16/CD32
Clone 2.4G2
Concentration 0.5mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15831279
View Documents
BD Biosciences
561728
0.1 mg
ONLINE EXCLUSIVE PRICE
£159.00
£151.60 / Each
Save £7.40
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15897028
View Documents
BD Biosciences
553144
0.5 mg
ONLINE EXCLUSIVE PRICE
£434.00
£417.13 / Each
Save £16.87
3% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The 2.4G2 antibody reacts specifically with a common nonpolymorphic epitope on the extracellular domains of the mouse FcγIII and FcγII receptors. It has also been reported to bind the FcγI receptor (CD64) via its Fc domain. 2.4G2 mAb blocks non-antigen-specific binding of immunoglobulins to the FcγIII and FcγII, and possibly FcγI, receptors in vitro and in vivo. CD16 and/or CD32 are expressed on natural killer cells, monocytes, macrophages, dendritic cells (at low levels), Kupffer cells, granulocytes, mast cells, B lymphocytes, immature thymocytes, and some activated mature T lymphocytes.
Specifications

Specifications

CD16/CD32
0.5mg/mL
Monoclonal
Rat
Mouse
Mouse BALB/c Macrophage J774
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
2.4G2
Flow Cytometry
FITC
RUO
Aqueous buffered solution containing ≤0.09% sodium azide.
IgG2b κ
Affinity Purified
FcγRIII/FcγRII; Fcgr3/Fcgr2
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.