Learn More
CD158A Mouse anti-Human, Brilliant Violet 711, Clone: HP-3E4, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 563183
Description
The HP-3E4 monoclonal antibody reacts with CD158a. CD158a is a 58 kDa membrane glycoprotein, member of the killer-inhibitory receptor (KIR). The KIR family family consists of transmembrane glycoproteins of the Ig superfamily expressed on <1-8% of NK cells and a subset of T cells which are involved in recognition of MHC class I molecules on target cells and inhibit cytotoxicity. CD158a is an antigen which recognizes HLA-C molecules (-Cw*0401 and -Cw*1503). This antibody is suitable for staining acetone-fixed, frozen tissue sections. It was reported that this clone also crossreacted with NKAT8 (CD158i) tranfectant cells.
The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. This dye is a tandem fluorochrome of BD Horizon™ BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 711-nm. BD Horizon™ BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy™5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination.
Specifications
| CD158a | |
| Monoclonal | |
| Brilliant Violet 711 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human NK Cells | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| HP-3E4 | |
| KIR2DL1; KI2L1; NKAT; NKAT-1; NKAT1; KIR-K64; KIR221; p58.1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgM |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.