Learn More
CD152 Mouse anti-Human, APC, Clone: BNI3, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 560938
Description
Reacts with the “cytolytic T lymphocyte-associated antigen”, CTLA-4. CTLA-4 is transiently expressed on activated CD28+ T cells and binds to CD80 and CD86 present on antigen presenting cells (APC) with high avidity. This interaction appears to deliver a negative regulatory signal to the T cell. There are recent reports that indicate that CTLA-4 is also expressed on B cells when cultured with activated T cells, suggesting a possible role of CTLA-4 in the regulation of B-cell response. Immobilized BNI3.1 enhances T-cell proliferation induced by CD3 and CD28. Recent studies have showed that CD152 can be expressed by regulatory T (Treg) cells. It has been found this antibody can stain the
intracellular CD152 on the Treg cells after fixation and permeabilization of cells.
Host Species: Mouse
Clone: BNI3
Isotype: IgG2a κ
Species Reactivity: Human
Flow Cytometry, Intracellular Staining
Specifications
| CD152 | |
| Monoclonal | |
| APC | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human CTLA4 Recombinant Protein | |
| 25 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| BNI3 | |
| CTLA-4; AILIM; Cytotoxic T-lymphocyte protein 4 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.