Learn More
CD15 Mouse anti-Human, Unlabeled, Clone: MMA, BD
Mouse Monoclonal Antibody
Brand:  BD Biosciences 347420
Product Code. 15816098
  • £280.00 / Each
Description
CD15 recognizes a human myelomonocytic antigen. The structure recognized by CD15 antibodies is lacto-N-fucopentose III.The CD15 antigen is present on greater than 95% of mature peripheral blood eosinophils and neutrophils and is present at low density on circulating monocytes.In lymphoid tissue, CD15 reacts with Reed-Sternberg cells of Hodgkin's disease and with granulocytes. However, CD15 reacts with few tissue macrophages and does not react with dendritic cells.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD15
Monoclonal
50μg/mL
HEPES buffer with gelatin and 0.1% sodium azide.
U-937 histiocytic cell line
100 Tests
Cell Biology
Human
IgM κ
Flow Cytometry
MMA
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.