Learn More
CD144 Mouse anti-Human, PerCP-Cy5.5, Clone: 55-7H1, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 561566
Description
The 55-7H1 antibody reacts with a calcium-independent epitope on cadherin-5, a member of the cadherin family of calcium-dependent adhesion molecules. Cadherin-5 is expressed on endothelial cells in vivo and in vitro. It may play a role in the organization of lateral endothelial junctions and in the control of permeability properties of vascular endothelium.
Specifications
| CD144 | |
| Monoclonal | |
| PerCP-Cyanine5.5 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| 55-7H1 | |
| VE-cadherin; Cadherin-5; CDH5; Vascular endothelial cadherin | |
| Mouse | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.