Learn More
CD14 Mouse anti-Human, BD IMag-DM, Clone: MΦP9, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 557769
Description
BD IMag™ Anti-Human CD14 Magnetic Particles - DM are magnetic nanoparticles that have monoclonal antibody conjugated to their surfaces. These particles are optimized for the positive selection or depletion of CD14-bearing leukocytes using the BD IMagnet™. CD14 has been reported to be expressed at high levels on peripheral blood monocytes. Additionally, it is found on interfollicular macrophages, reticular dendritic cells, and some Langerhans cells. BD IMag™ Anti-Human CD14 magnetic particles have been reported to effectively separate the CD14-bearing cells of rhesus macaque blood.
Peripheral Blood Mononuclear Cells (PBMC) are labeled with BD IMag™ Anti-human CD14 Magnetic Particles - DM according to the Magnetic Labeling Protocol. This labeled cell suspension is then placed within the magnetic field of the BD IMagnet™ (Cat. no. 552311). Labeled cells migrate toward the magnet (positive fraction), leaving the unlabeled cells in suspension so they can be drawn off (negative fraction). The tube is then removed from the magnetic field for resuspension of the positive fraction. The separation is repeated twice to increase the purity of the positive fraction. The magnetic separation steps are diagrammed in the Separation Flow Chart. After the positive fraction is washed, the small size of the magnetic particles allows the positive fraction to be further evaluated in downstream applications such as flow cytometry.
Specifications
| CD14 | |
| Monoclonal | |
| BD IMag-DM | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human Monocytes | |
| 1 x 109 Cells | |
| Immunology | |
| Anti-Human CD14 Magnetic Particles, Mouse (BALB/c) IgG2b, κ (5mL) | |
| Store undiluted at 4°C |
| Flow Cytometry | |
| MΦP9 | |
| LPS receptor; LPS-R; Myeloid cell-specific leucine-rich glycoprotein | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Rhesus Monkey | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.